Methyl-2-fluoro-4-hydroxybenzoate ≥95%
Supplier: Matrix Scientific
074868-1G
074868-5G
102538-608EA
64.62
USD
102538-608
102538-610
Methyl-2-fluoro-4-hydroxybenzoate ≥95%
Methyl-2-fluoro-4-hydroxybenzoate
Hazard: Irritant
Formula:
C₈H₇FO₃ |
MDL Number:
MFCD09038282 CAS Number: 197507-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...