4-Bromo-5-thiazolecarboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

074866-250MG 074866-1G
102538-600EA 967.69 USD
102538-600 102538-602
4-Bromo-5-thiazolecarboxylic acid ethyl ester ≥95%
4-Bromo-5-thiazolecarboxylic acid ethyl ester

Hazard: Irritant

Formula: C₆H₆BrNO₂S
MDL Number: MFCD09909677
CAS Number: 152300-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR