5,6,12,13-Tetrachloroanthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone ≥95%

Supplier: Matrix Scientific

074865-1G 074865-5G
102538-596EA 284.62 USD
102538-596 102538-598
5,6,12,13-Tetrachloroanthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone ≥95%
5,6,12,13-Tetrachloroanthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone

Hazard: Irritant

Formula: C₂₄H₄Cl₄O₆
MDL Number: MFCD08056065
CAS Number: 156028-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR