3-Nitro-2,6-lutidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dimethyl-3-nitropyridine, 3-Nitro-2,6-lutidine

074864-1G 074864-5G 074864-10G
102538-590EA 72.31 USD
102538-590 102538-592 102538-594
3-Nitro-2,6-lutidine ≥95%
3-Nitro-2,6-lutidine

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD00023503
CAS Number: 15513-52-7
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR