Sodium-2-naphthol-6-sulfonate hydrate ≥95%

Supplier: Matrix Scientific

074856-10G 074856-100G
102538-562EA 55.38 USD
102538-562 102538-564
Sodium-2-naphthol-6-sulfonate hydrate ≥95%
Sodium-2-naphthol-6-sulfonate hydrate

Hazard: Irritant

Formula: C₁₀H₈O₄S
MDL Number: MFCD00070488
CAS Number: 135-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR