H-Asn(Trt)-OH ≥95%

Supplier: Matrix Scientific

Synonyms: Nγ-Trityl-L-Asparagine, Ngamma-Trityl-L-asparagine, (S)-2-Amino-4-oxo-4-(tritylamino)butyric acid

074854-1G 074854-5G 074854-10G
102538-552EA 233.85 USD
102538-552 102538-554 102538-556
H-Asn(Trt)-OH ≥95%
H-Asn(Trt)-OH

Hazard: Irritant

Formula: C₂₃H₂₂N₂O₃
MDL Number: MFCD00153422
CAS Number: 132388-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR