(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-4-phenyl-D-Phe-OH, N-Boc-3-(4-biphenylyl)-D-alanine

074852-1G 074852-5G 074852-10G
102538-540EA 67.69 USD
102538-540 102538-542 102538-544
(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥95%
(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

Hazard: Irritant

Formula: C₂₀H₂₃NO₄
MW: 341.41 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00191185
CAS Number: 128779-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR