N-Boc-4-oxo-L-proline ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-4-oxo-Pro-OH, (S)-1-Boc-4-oxo-2-pyrrolidinecarboxylic Acid, (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, (S)-1-(tert-Butoxycarbonyl)-4-oxo-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4-oxo-L-proline

074850-5G 074850-25G
102538-530EA 153.85 USD
102538-530 102833-730
N-Boc-4-oxo-L-proline ≥95%
N-Boc-4-oxo-L-proline

Hazard: Irritant

Formula: C₁₀H₁₅NO₅
MW: 229.23 g/mol
MDL Number: MFCD01860669
CAS Number: 84348-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR