4-(Methylcarbamoyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Methylcarbamoyl)benzeneboronic acid, 4-(N-Methylaminocarbonyl)phenylboronic acid

074848-1G 074848-5G
102538-520EA 203.08 USD
102538-520 102538-522
4-(Methylcarbamoyl)phenylboronic acid ≥95%
4-(Methylcarbamoyl)phenylboronic acid

Hazard: Irritant

Formula: C₈H₁₀BNO₃
MDL Number: MFCD03788427
CAS Number: 121177-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR