4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)-2-morpholinyl)methyl)benzamide ≥95%

Supplier: Matrix Scientific

Synonyms: Mosapride, 4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)morpholin-2-yl)methyl)benzamide

074845-1G 074845-5G
102538-508EA 569.23 USD
102538-508 102538-510
4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)-2-morpholinyl)methyl)benzamide ≥95%
4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)-2-morpholinyl)methyl)benzamide

Hazard: Irritant

Formula: C₂₁H₂₅ClFN₃O₃
MW: 421.89 g/mol
MDL Number: MFCD00867430
CAS Number: 112885-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR