(±)-2-Methylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methylpiperazine

074843-100G 074843-500G
102538-502EA 184.62 USD
102538-502 102833-728
(±)-2-Methylpiperazine ≥95%
(±)-2-Methylpiperazine

Hazard: Flammable Solid/Hygroscopic

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 155°C/763mmHg
Melting Pt: 61-63°C
Flash Pt: 65°C
MDL Number: MFCD00005954
CAS Number: 109-07-9
UN: 2925
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR