(S)-(+)-β-Aminophenethyl alcohol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

074841-100G 074841-25G
102538-496EA 135.38 USD
102538-496 102833-726
(S)-(+)-β-Aminophenethyl alcohol ≥95%
(S)-(+)-β-Aminophenethyl alcohol

Hazard: Irritant/Air Sensitive

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD00064404
CAS Number: 20989-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR