(1S,2R)-2-(Benzyloxymethyl)-1-hydroxy-3-cyclopentene ≥90%

Supplier: Matrix Scientific

074837-1G 074837-5G
102538-476EA 229.23 USD
102538-476 102538-478
(1S,2R)-2-(Benzyloxymethyl)-1-hydroxy-3-cyclopentene ≥90%
(1S,2R)-2-(Benzyloxymethyl)-1-hydroxy-3-cyclopentene

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MDL Number: MFCD09263486
CAS Number: 110567-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR