Methyl-3-phenylpropionate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl dihydrocinnamate, 3-Phenylpropionic acid methyl ester, Methyl 3-phenylpropionate, Hydrocinnamic acid methyl ester

074836-100G 074836-500G
102538-470EA 320 USD
102538-470 102538-472
Methyl-3-phenylpropionate ≥95%
Methyl-3-phenylpropionate

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00017209
CAS Number: 103-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR