Methyl-1-benzylpiperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

074834-10G 074834-100G
102538-464EA 46.15 USD
102538-464 102538-466
Methyl-1-benzylpiperidine-4-carboxylate ≥95%
Methyl-1-benzylpiperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₂
MDL Number: MFCD09750949
CAS Number: 10315-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR