7-Bromo-1-heptanol ≥95%

Supplier: Matrix Scientific

074832-1G 074832-5G
102538-454EA 84.62 USD
102538-454 102538-456
7-Bromo-1-heptanol ≥95%
7-Bromo-1-heptanol

Hazard: Irritant

Formula: C₇H₁₅BrO
MW: 195.10 g/mol
Boiling Pt: 11-112°C/4mm
Density: 1.269
MDL Number: MFCD00041708
CAS Number: 10160-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR