Benzyltrimethylammonium hydroxide 40% in methanol , 95%

Supplier: Matrix Scientific

Synonyms: Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide

074831-10G 074831-100G
102538-450EA 36.92 USD
102538-450 102538-452
Benzyltrimethylammonium hydroxide 40% in methanol , 95%
Benzyltrimethylammonium hydroxide

Hazard: Irritant

Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
MDL Number: MFCD00008281
CAS Number: 100-85-6
UN: 3286
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR