Pyridine-2,5-dicarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Isocinchomeronic acid, 2,5-Pyridinedicarboxylic acid

074829-100G 074829-25G
102538-444EA 135.38 USD
102538-444 102833-720
Pyridine-2,5-dicarboxylic acid ≥95%
Pyridine-2,5-dicarboxylic acid

Hazard: Irritant

Formula: C₇H₅NO₄
MW: 167.12 g/mol
MDL Number: MFCD00006297
CAS Number: 100-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR