Bis-trispropane ≥95%

Supplier: Matrix Scientific

074826-10G 074826-100G
102538-428EA 161.54 USD
102538-428 102538-430
Bis-trispropane ≥95%
Bis-trispropane

Hazard: Irritant

Formula: C₁₁H₂₆N₂O₆
MDL Number: MFCD00004689
CAS Number: 64431-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR