4-Morpholin-4-yl-2-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

074824-1G 074824-5G
102538-420EA 533.85 USD
102538-420 102538-422
4-Morpholin-4-yl-2-nitrobenzoic acid ≥95%
4-Morpholin-4-yl-2-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₅
MW: 252.2200 g/mol
MDL Number: MFCD12198419
CAS Number: 404010-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR