(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
Boc-Tyr(tBu)-OH, N-Boc-O-tert-butyl-L-tyrosine
074822-1G
074822-5G
074822-10G
102538-410EA
55.38
USD
102538-410
102538-412
102538-414
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥95%
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
Hazard: Irritant
Formula:
C₁₈H₂₇NO₅ MW: 337.4 g/mol |
MDL Number:
MFCD00065598 CAS Number: 47375-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...