5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-3-(trifluoromethyl)pyridin-2-ol
074821-1G
074821-5G
102538-404EA
158.46
USD
102538-404
102538-406
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine ≥95%
5-Bromo-2-hydroxy-3-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₃BrF₃NO MW: 242.00 g/mol |
MDL Number:
MFCD12546486 CAS Number: 76041-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...