D-Norvaline ≥95%

Supplier: Matrix Scientific

Synonyms: H-D-Nva-OH, (R)-(-)-Norvaline, D(-)-Norvaline

074819-10G 074819-100G 074819-500G
102538-394EA 126.15 USD
102538-394 102538-396 102538-398
D-Norvaline ≥95%
D-Norvaline

Hazard: Irritant

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 222-223°C
MDL Number: MFCD00008097
CAS Number: 2013-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR