Piperidin-1-ium acetate ≥95%

Supplier: Matrix Scientific

074818-100G 074818-25G
102538-392EA 576.92 USD
102538-392 102833-718
Piperidin-1-ium acetate ≥95%
Piperidin-1-ium acetate

Hazard: Irritant

Formula: C₇H₁₅NO₂
CAS Number: 4540-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR