4-(2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-4-morpholinophenylboronic Acid Pinacol Ester, 3-Chloro-4-(4-morpholinyl)benzeneboronic acid pinacol ester, 4-[2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-[2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
074817-1G
074817-5G
102538-386EA
640
USD
102538-386
102538-388
4-(2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine ≥95%
4-(2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine
Hazard: Irritant
Formula:
C₁₆H₂₃BClNO₃ MW: 323.63 g/mol |
MDL Number:
MFCD18070979 CAS Number: 1361110-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...