2-Bromo-4-methoxybenzonitrile ≥95%

Supplier: Matrix Scientific

074816-1G 074816-5G
102538-380EA 36.92 USD
102538-380 102538-382
2-Bromo-4-methoxybenzonitrile ≥95%
2-Bromo-4-methoxybenzonitrile

Hazard: Irritant

Formula: C₈H₆BrNO
MW: 212.0433 g/mol
MDL Number: MFCD09056764
CAS Number: 140860-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR