6-Bromo-1,2-benzisoxazole ≥95%

Supplier: Matrix Scientific

074815-250MG 074815-1G 074815-5G
102538-374EA 200 USD
102538-374 102538-376 102538-378
6-Bromo-1,2-benzisoxazole ≥95%
6-Bromo-1,2-benzisoxazole

Hazard: Irritant

Formula: C₇H₄BrNO
MW: 198.0168 g/mol
MDL Number: MFCD06659634
CAS Number: 1060802-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR