Benzothiazole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

074813-1G 074813-5G
102538-364EA 240 USD
102538-364 102538-366
Benzothiazole-2-carboxylic acid ≥95%
Benzothiazole-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₅NO₂S
Melting Pt: 106 °C (dec.)
MDL Number: MFCD02682025
CAS Number: 3622-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR