Boc-D-valinol ≥95%

Supplier: Matrix Scientific

Synonyms: N-BOC-D-Valinol, Boc-D-Valinol

074811-1G 074811-5G 074811-10G
102538-352EA 107.69 USD
102538-352 102538-354 102538-356
Boc-D-valinol ≥95%
Boc-D-valinol

Hazard: Irritant

Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
MDL Number: MFCD00235960
CAS Number: 106391-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR