1-(2-Phenoxyphenyl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

074810-250MG 074810-1G 074810-5G
102538-346EA 456.92 USD
102538-346 102538-348 102538-350
1-(2-Phenoxyphenyl)methanamine hydrochloride ≥95%
1-(2-Phenoxyphenyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₄ClNO
MW: 235.7094 g/mol
MDL Number: MFCD08271962
CAS Number: 31963-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR