6-Nitro-o-cresol ≥95%
Supplier: Matrix Scientific
074808-1G
074808-5G
074808-10G
102538-334EA
30.77
USD
102538-334
102538-336
102538-338
6-Nitro-o-cresol ≥95%
6-Nitro-o-cresol
Hazard: Irritant
Formula:
C₇H₇NO₃ |
MDL Number:
MFCD00031116 CAS Number: 13073-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...