5-Nitro-o-cresol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Hydroxy-2-methyl-5-nitrobenzene, 2-Methyl-5-nitrophenol

074806-5G 074806-10G 074806-100G
102538-322EA 15.38 USD
102538-322 102538-324 102538-326
5-Nitro-o-cresol ≥95%
5-Nitro-o-cresol

Hazard: Toxic

Formula: C₇H₇NO₃
Melting Pt: 116-120°C
MDL Number: MFCD00043909
CAS Number: 5428-54-6
UN: 2446
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR