(2R,4R)-4-Methylpiperidine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

074804-250MG 074804-1G 074804-5G
102538-312EA 483.85 USD
102538-312 102538-314 102538-316
(2R,4R)-4-Methylpiperidine-2-carboxylic acid ≥95%
(2R,4R)-4-Methylpiperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₃NO₂
MW: 143.1800 g/mol
MDL Number: MFCD03839858
CAS Number: 74892-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR