(R)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine ≥95%

Supplier: Matrix Scientific

074802-250MG 074802-1G 074802-5G
102538-300EA 215.38 USD
102538-300 102538-302 102538-304
(R)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine ≥95%
(R)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂
CAS Number: 155726-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR