Methyl-3-chloro-2-hydroxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-chloro-2-hydroxybenzoate

074799-250MG 074799-1G 074799-5G
102538-282EA 283.08 USD
102538-282 102538-284 102538-286
Methyl-3-chloro-2-hydroxybenzoate ≥95%
Methyl-3-chloro-2-hydroxybenzoate

Hazard: Irritant

Formula: C₈H₇ClO₃
MW: 186.5924 g/mol
MDL Number: MFCD00128116
CAS Number: 52159-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR