6-Bromo-3-iodoimidazo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

074798-1G 074798-5G 074798-10G
102538-276EA 227.69 USD
102538-276 102538-278 102538-280
6-Bromo-3-iodoimidazo[1,2-a]pyrazine ≥95%
6-Bromo-3-iodoimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₆H₃BrIN₃
MDL Number: MFCD18072708
CAS Number: 1245644-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR