3-Bromo-6-mercaptopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromopyridine-2-thiol

074796-1G 074796-5G
102538-266EA 110.77 USD
102538-266 102538-268
3-Bromo-6-mercaptopyridine ≥95%
3-Bromo-6-mercaptopyridine

Hazard: Irritant

Formula: C5H4BrNS
MW: 190.0600 g/mol
MDL Number: MFCD09729468
CAS Number: 56673-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR