Methyl-2-methylthiophene-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-methylthiophene-3-carboxylate

074792-1G 074792-5G
102538-244EA 269.23 USD
102538-244 102538-246
Methyl-2-methylthiophene-3-carboxylate ≥95%
Methyl-2-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₇H₈O₂S
MW: 156.2000 g/mol
MDL Number: MFCD12026360
CAS Number: 53562-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR