2,6-Difluoro-4-methoxyphenylboronic acid ≥95%

Supplier: Matrix Scientific

074791-1G 074791-5G
102538-240EA 90.77 USD
102538-240 102538-242
2,6-Difluoro-4-methoxyphenylboronic acid ≥95%
2,6-Difluoro-4-methoxyphenylboronic acid

Hazard: Irritant

Formula: C₇H₇BF₂O₃
MDL Number: MFCD05664299
CAS Number: 406482-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR