5-Chloro-1H-indole-3-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-3-cyanoindole

074789-250MG 074789-1G 074789-5G
102538-228EA 355.38 USD
102538-228 102538-230 102538-232
5-Chloro-1H-indole-3-carbonitrile ≥95%
5-Chloro-1H-indole-3-carbonitrile

Hazard: Irritant

Formula: C₉H₅ClN₂
MW: 176.61 g/mol
MDL Number: MFCD07776618
CAS Number: 194490-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR