4-Cyanobutanoic acid ≥95%

Supplier: Matrix Scientific

074786-250MG 074786-1G 074786-5G
102538-214EA 400 USD
102538-214 102538-216 102538-218
4-Cyanobutanoic acid ≥95%
4-Cyanobutanoic acid

Hazard: Irritant

Formula: C₅H₇NO₂
MW: 113.12 g/mol
MDL Number: MFCD00218781
CAS Number: 39201-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR