Methyl-5-bromothiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-thiophene-2-carboxylic acid methyl ester, Methyl 5-bromothiophene-2-carboxylate

074785-1G 074785-5G
102538-210EA 44.62 USD
102538-210 102538-212
Methyl-5-bromothiophene-2-carboxylate ≥95%
Methyl-5-bromothiophene-2-carboxylate

Hazard: Irritant

Formula: C₆H₅BrO₂S
MW: 221.0700 g/mol
MDL Number: MFCD03233845
CAS Number: 62224-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR