1-Bromo-2-fluoro-5-iodobenzene ≥95%

Supplier: Matrix Scientific

074783-5G 074783-100G
102538-198EA 96.92 USD
102538-198 102538-202
1-Bromo-2-fluoro-5-iodobenzene ≥95%
1-Bromo-2-fluoro-5-iodobenzene

Hazard: Irritant/Light Sensitive/Refrigerate

Formula: C₆H₃BrFI
MDL Number: MFCD04116322
CAS Number: 811842-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR