4-Picolyl bromide hydrobromide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)pyridine hydrobromide

074782-5G 074782-25G
102538-194EA 400 USD
102538-194 102833-716
4-Picolyl bromide hydrobromide ≥95%
4-Picolyl bromide hydrobromide

Hazard: Irritant

Formula: C₆H₇Br₂N
MW: 252.94 g/mol
MDL Number: MFCD01863545
CAS Number: 73870-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR