EMCA (6-Maleimidohexanoic acid) ≥95%

Supplier: Matrix Scientific

Synonyms: N-Maleoyl-6-aminocaproic acid, ε-Maleimidocaproic acid, 2,5-Dihydro-2,5-dioxo-1H-pyrrole-1-hexanoic acid, 6-Maleimide hexanoic acid, N-(5-Carboxypentyl)maleimide, MCA, Maleimidocaproic acid, 6-Maleimidohexanoic acid, 6-Maleimidocaproic acid

074777-1G 074777-5G
102538-168EA 38.46 USD
102538-168 102538-170
EMCA (6-Maleimidohexanoic acid) ≥95%
EMCA (6-Maleimidohexanoic acid)

Hazard: Irritant

Formula: C₁₀H₁₃NO₄
MW: 211.22 g/mol
MDL Number: MFCD00043140
CAS Number: 55750-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR