tert-Butyl carbamate ≥95%
Supplier: Matrix Scientific
Synonyms:
Carbamic acid tert-butyl ester
074774-100G
074774-25G
102538-158EA
73.85
USD
102538-158
102833-714
tert-Butyl carbamate ≥95%
tert-Butyl carbamate
Hazard: Irritant
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol |
MDL Number:
MFCD00007962 CAS Number: 4248-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...