2,4-Dimethylpiperidine hydrochloride ≥95%

Supplier: Matrix Scientific

074769-250MG 074769-1G
102538-132EA 498.46 USD
102538-132 102538-134
2,4-Dimethylpiperidine hydrochloride ≥95%
2,4-Dimethylpiperidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClN
CAS Number: 91846-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR