3'-Carbamoyl-[1,1'-biphenyl]-3-ylcyclohexylcarbamate ≥95%
Supplier: Matrix Scientific
Synonyms:
URB597
074768-1G
074768-5G
102538-128EA
710.77
USD
102538-128
102538-130
3'-Carbamoyl-[1,1'-biphenyl]-3-ylcyclohexylcarbamate ≥95%
3'-Carbamoyl-[1,1'-biphenyl]-3-ylcyclohexylcarbamate
Hazard: Irritant
Formula:
C20H22N2O3 MW: 338.4 g/mol |
CAS Number:
546141-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...