(1-(4-Nitrophenyl)piperidin-3-yl)methanol ≥95%
Supplier: Matrix Scientific
074766-1G
074766-5G
102538-118EA
640
USD
102538-118
102538-120
(1-(4-Nitrophenyl)piperidin-3-yl)methanol ≥95%
(1-(4-Nitrophenyl)piperidin-3-yl)methanol
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₃ |
MDL Number:
MFCD11039478 CAS Number: 166438-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...