N-Methylphenylalanine ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Me-Phe-OH
074765-1G
074765-5G
074765-10G
102538-112EA
61.54
USD
102538-112
102538-114
102538-116
N-Methylphenylalanine ≥95%
N-Methylphenylalanine
Hazard: Irritant
Formula:
C₁₀H₁₃NO₂ MW: 179.219 g/mol |
MDL Number:
MFCD00037757 CAS Number: 2566-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...